siRNA Details
Gene Symbol: Gm36455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041461.1 | - | 2 | 283 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638138.1 | - | 2 | 279 | Chr. 16 - 21834080 - 21836554 [-] on Build GRCm38 |
