siRNA Details
Gene Symbol: 4930509J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040547.1 | 4 | 290 | |||
GenBank mRNA | AK015737.1 | - | 4 | 292 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715278.1 | - | 4 | 292 | Chr. 3 - 73362027 - 73481651 [-] on Build GRCm38 |
