siRNA Details
Gene Symbol: 4930509K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040663.1 | 1 | 161 | |||
GenBank mRNA | AK015738.1 | - | 1 | 162 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715279.1 | - | 1 | 162 | Chr. 4 - 40313232 - 40316868 [+] on Build GRCm38 |
