siRNA Details
Gene Symbol: Gm13016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043727.1 | - | 3 | 403 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB641446.1 | - | 3 | 401 | Chr. 4 - 140380631 - 140384533 [+] on Build GRCm38 |
