siRNA Details
Gene Symbol: 5830453J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018006.1 | - | 1 | 314 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717937.1 | - | 1 | 314 | Chr. 11 - 46377713 - 46378581 [-] on Build GRCm38 |
