siRNA Details
Gene Symbol: Gm35066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035404.1 | - | 2 | 296 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627748.1 | - | 2 | 296 | Chr. 3 - 144972124 - 144981228 [+] on Build GRCm38 |
