siRNA Details
Gene Symbol: Gm29154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131742.1 | - | 4 | 366 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706784.1 | - | 4 | 366 | Chr. 9 - 103919501 - 103959421 [-] on Build GRCm38 |
