siRNA Details
Gene Symbol: 1700016L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006046.1 | - | 1 | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706162.1 | - | 1 | 233 | Chr. 7 - 131937874 - 131938127 [+] on Build GRCm38 |
