siRNA Details
Gene Symbol: 1700016L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040460.1 | 3 | 164 | |||
GenBank mRNA | AK006047.1 | - | 3 | 165 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706163.1 | - | 3 | 162 | Chr. 1 - 80445934 - 80475660 [+] on Build GRCm38 |
