siRNA Details
Gene Symbol: Gm31463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053231.1 | - | 2 | 342 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664580.1 | - | 2 | 337 | Chr. 8 - 8576528 - 8578638 [+] on Build GRCm38 |
