siRNA Details
Gene Symbol: Miat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003718.2 | 7 | 3283 | |||
NR_033657.1 | 7 | 3327 | ||||
GenBank mRNA | AB300594.1 | - | 7 | 3270 | ||
AK053540.1 | - | 7 | 3273 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Miat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003718.2 | 7 | 3283 | |||
NR_033657.1 | 7 | 3327 | ||||
GenBank mRNA | AB300594.1 | - | 7 | 3270 | ||
AK053540.1 | - | 7 | 3273 |