siRNA Details
Gene Symbol: Miat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053540.1 | - | 1, 2 | 328 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB665139.1 | - | 1, 2 | 325 | Chr. 5 - 112224233 - 112228933 [-] on Build GRCm38 |
