siRNA Details
Gene Symbol: Gm33035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135793.1 | - | 3 | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ052621.1 | - | 3 | 395 | Chr. 10 - 69021812 - 69033888 [-] on Build GRCm38 |
