siRNA Details
Gene Symbol: Gm19325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135790.1 | - | 921 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB700210.1 | - | 2 | 416 | Chr. 9 - 97047444 - 97048333 [-] on Build GRCm38 |
