siRNA Details
Gene Symbol: Gm15350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045775.1 | 3 | 472 | |||
GenBank mRNA | AK006628.1 | - | 3 | 474 | ||
AK018911.1 | - | 3 | 473 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707073.1 | - | 3 | 473 | Chr. 8 - 12828335 - 12831919 [-] on Build GRCm38 |
