siRNA Details
Gene Symbol: Gm9925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036329.1 | - | 2 | 1978 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB132020.2 | - | 2 | 517 | Chr. 18 - 74066564 - 74068512 [+] on Build GRCm38 |
