siRNA Details
Gene Symbol: C030044M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021148.1 | - | 1 | 1190 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB360048.1 | - | 1 | 260 | Chr. 7 - 16147123 - 16147387 [+] on Build GRCm38 |
