siRNA Details
Gene Symbol: Gm15866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079714.1 | - | 5 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB200474.2 | - | 2 | 438 | Chr. 5 - 43557289 - 43599223 [-] on Build GRCm38 |
