siRNA Details
Gene Symbol: Gm1653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040591.1 | 1 | 382 | |||
GenBank mRNA | AK133793.1 | - | 1 | 383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB618725.1 | - | 1 | 383 | Chr. 3 - 149274737 - 149278199 [+] on Build GRCm38 |
