siRNA Details
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076483.1 | - | 1 | 1350 | ||
BC048860.1 | - | 1 | 2537 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076483.1 | - | 1 | 1350 | ||
BC048860.1 | - | 1 | 2537 |