siRNA Details
Gene Symbol: Gm19939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045942.1 | - | 1 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DW712250.1 | - | 1 | 414 | Chr. 18 - 51131280 - 51131876 [+] on Build GRCm38 |
