siRNA Details
Gene Symbol: Gm11981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046025.1 | 3 | 292 | |||
GenBank mRNA | AK076664.1 | - | 3 | 293 | ||
BB614940.1 | - | 3 | 293 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV260002.2 | - | 3 | 262 | Chr. 11 - 6715999 - 6720006 [-] on Build GRCm38 |
