siRNA Details
Gene Symbol: Gm40466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029792.1 | - | 2 | 3806 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB016866.2 | - | 1 | 230 | Chr. 8 - 20004353 - 20005008 [+] on Build GRCm38 |
