siRNA Details
Gene Symbol: 2610011E03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011375.1 | - | 1 | 2094 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV112314.1 | - | 1 | 94 | Chr. 5 - 131996183 - 131996411 [-] on Build GRCm38 |
