siRNA Details
Gene Symbol: 1700047N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006719.1 | - | 1 | 457 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706708.1 | - | 1 | 457 | Chr. 1 - 10057057 - 10057944 [-] on Build GRCm38 |
