siRNA Details
Gene Symbol: 1700119I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018965.1 | - | 2 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707296.1 | - | 2 | 172 | Chr. 13 - 91861432 - 91862056 [+] on Build GRCm38 |
