siRNA Details
Gene Symbol: Gm41341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144172.1 | - | 1 | 34 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY733079.1 | - | 1 | 33 | Chr. 15 - 68878315 - 68883391 [+] on Build GRCm38 |
