siRNA Details
Gene Symbol: Gm29811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102297.1 | 4 | 1033 | |||
NR_102298.1 | 4 | 1030 | ||||
GenBank mRNA | AK135894.1 | - | 4 | 1034 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CO799428.1 | - | 4 | 560 | Chr. 3 - 132842720 - 132878120 [-] on Build GRCm38 |
