siRNA Details
Gene Symbol: 9130016M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018631.1 | - | 1 | 594 | ||
AK050221.1 | - | 1 | 1739 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720812.1 | - | 1 | 594 | Chr. 19 - 31951227 - 31952194 [+] on Build GRCm38 |
