siRNA Details
Gene Symbol: 4930447E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019616.1 | - | 1 | 560 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714955.1 | - | 1 | 560 | Chr. 5 - 101032634 - 101033252 [-] on Build GRCm38 |
