siRNA Details
Gene Symbol: 5830416P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019949.1 | - | 2 | 217 | ||
AK042769.1 | - | 2 | 177 | |||
AK047177.1 | - | 2 | 171 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5830416P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019949.1 | - | 2 | 217 | ||
AK042769.1 | - | 2 | 177 | |||
AK047177.1 | - | 2 | 171 |