siRNA Details
Gene Symbol: 1700066J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006903.1 | - | 1 | 26 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706900.1 | - | 1 | 22 | Chr. 13 - 58146952 - 58148974 [+] on Build GRCm38 |
