siRNA Details
Gene Symbol: D430006K04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084893.1 | - | 2 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB601103.2 | - | 2 | 500 | Chr. 17 - 78882042 - 78882919 [+] on Build GRCm38 |
