siRNA Details
Gene Symbol: Nkx1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF222442.1 | - | 1452 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV308505.2 | - | 1 | 483 | Chr. 7 - 132594882 - 132595624 [-] on Build GRCm38 |
