siRNA Details
Gene Symbol: BC042761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042761.1 | - | 1 | 274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF534285.1 | - | 1 | 274 | Chr. 12 - 57661458 - 57662911 [-] on Build GRCm38 |
