siRNA Details
Gene Symbol: C030013C02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021071.1 | - | 1 | 103 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728150.1 | - | 1 | 99 | Chr. 6 - 80504878 - 80505358 [+] on Build GRCm38 |
