siRNA Details
Gene Symbol: Dmrta2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019375.1 | - | 1 | 100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712842.1 | - | 1 | 100 | Chr. 4 - 109976744 - 109977491 [-] on Build GRCm38 |
