siRNA Details
Gene Symbol: 4930429L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015242.1 | - | 1 | 276 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714799.1 | - | 1 | 276 | Chr. 4 - 151557365 - 151558273 [+] on Build GRCm38 |
