siRNA Details
Gene Symbol: 4930448L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019618.1 | - | 1 | 260 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714969.1 | - | 1 | 260 | Chr. 11 - 30771290 - 30771674 [-] on Build GRCm38 |
