siRNA Details
Gene Symbol: C030009J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046979.1 | - | 1 | 51 | ||
AK047676.1 | - | 1 | 45 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB647631.1 | - | 1 | 45 | Chr. 12 - 102270396 - 102271032 [+] on Build GRCm38 |
