siRNA Details
Gene Symbol: Trp53cor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036469.2 | 2 | 2637 | |||
GenBank mRNA | AK144811.1 | - | 2 | 2473 | ||
HM210889.1 | - | 3 | 2589 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trp53cor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036469.2 | 2 | 2637 | |||
GenBank mRNA | AK144811.1 | - | 2 | 2473 | ||
HM210889.1 | - | 3 | 2589 |