siRNA Details
Gene Symbol: 1700062C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006860.1 | - | 2 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706847.1 | - | 2 | 290 | Chr. 14 - 16000798 - 16037268 [-] on Build GRCm38 |
