siRNA Details
Gene Symbol: 4930451G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029584.1 | - | 1 | 2241 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV259418.2 | - | 1 | 165 | Chr. 4 - 141654245 - 141654917 [+] on Build GRCm38 |
