siRNA Details
Gene Symbol: 1700113H21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007196.1 | - | 1 | 114 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707284.1 | - | 1 | 114 | Chr. 1 - 20714277 - 20714743 [+] on Build GRCm38 |
