siRNA Details
Gene Symbol: 1700113B19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007192.1 | - | 1 | 249 | ||
AV209165.1 | - | 1 | 168 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707278.1 | - | 1 | 247 | Chr. 1 - 138261648 - 138262063 [+] on Build GRCm38 |
