siRNA Details
Gene Symbol: 4930584F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015867.1 | - | 3 | 562 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715418.1 | - | 3 | 562 | Chr. 5 - 26474935 - 26493289 [-] on Build GRCm38 |
