siRNA Details
Gene Symbol: 4921515E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045711.1 | 10 | 1975 | |||
GenBank mRNA | AK132982.1 | - | 10 | 1976 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV269390.2 | - | 3 | 342 | Chr. 15 - 18078142 - 18165866 [-] on Build GRCm38 |
