siRNA Details
Gene Symbol: 2900001A22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013458.1 | - | 1 | 160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV149847.1 | - | 1 | 50 | Chr. 7 - 138561894 - 138562184 [-] on Build GRCm38 |
