siRNA Details
Gene Symbol: Gm13964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131757.1 | - | 3 | 553 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707051.1 | - | 3 | 548 | Chr. 2 - 113500751 - 113504031 [-] on Build GRCm38 |
