siRNA Details
Gene Symbol: 8430413D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018407.1 | - | 1 | 356 | ||
AK048170.1 | - | 1 | 1368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720530.1 | - | 1 | 356 | Chr. 14 - 117204804 - 117205599 [+] on Build GRCm38 |
